Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methotrexate Antibody, Novus Biologicals™

Sheep Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Methotrexate |
---|---|
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017763
![]() |
Novus Biologicals
NB100659970.05ML |
0.05 mL |
Each for $181.90
|
|
|||||
NB10065997
![]() |
Novus Biologicals
NB10065997 |
0.25 mL |
Each for $487.50
|
|
|||||
Description
Methotrexate Polyclonal specifically detects Methotrexate in Non-species specific samples. It is validated for ELISA.Specifications
Methotrexate | |
ELISA | |
Unconjugated | |
Sheep | |
Non-species specific | |
MTX | |
IgG | |
Unpurified | |
NB100-65997 is specific for Methotrexate and exhibits the following cross reactivities with related compounds: Aminopterin (18%), Folic acid (<0.01%), Dyhydrofolic acid (<0.01%), Tetrahydrofolic acid (<0.01%), N-5-Methyltetrahydrofolic acid (<0.01%), Folinic acid (<0.01%), N-(p-Aminobenzoyl)-L-glutamic acid (<0.01%). |
ELISA 1:100-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with 0.09% Sodium Azide | |
Methotrexate (4-amino-10-methylpteroyl-L-glutamic acid) conjugated via the carboxyl groups of the glutamic acid residue to KLH. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title