Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methotrexate Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533142
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533142 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533142 Supplier Thermo Scientific Supplier No. PA533142
Add to Cart
Add to Cart

Methotrexate Polyclonal Antibody

Methotrexate Polyclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Methotrexate
Applications ELISA
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with <0.1% sodium azide
Gene Alias Abitrexate; Amethopterin; C20H22N8O5; Ledertrexate; Rheumatrex
Host Species Sheep
Immunogen Methotrexate (4-amino-10-methylpteroyl-L-glutamic acid) conjugated via the carboxyl groups of the glutamic acid residue to KLH.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.