Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methotrexate, PROMO, Thermo Scientific Chemicals

Catalog No. J6307503PM
Click to view available options
Quantity:
1 g

Solubility

Soluble in dilute HCl or dilute alkali hydroxide solutions (i.e. 0.1 M NaOH). Insoluble in plain water.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents. Protect from light

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.45 g/mol
MDL Number MFCD00150847
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Color Yellow
Quantity 1 g
UN Number UN1544
Sensitivity Hygroscopic; Light Sensitive
Merck Index 14,5985
Solubility Information Soluble in dilute HCl or dilute alkali hydroxide solutions ; Insoluble in plain water.
Physical Form Powder
Chemical Name or Material Methotrexate
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber 200-413-8
RTECSNumber MA1225000
TSCA Yes
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.