Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Alfa Aesar™

Light, Air & Moisture Sensitive

Manufacturer:  Alfa Aesar 4496703

Quantity 1g

 View more versions of this product

Catalog No. AA4496703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12148-71-9
Ir nominally 58%
662.88
1, 5-Cyclooctadiene(methoxy)iridium(I) dimer
Powder
C18H30Ir2O2
MFCD08459360
Methoxy(cyclooctadiene)iridium(I) dimer

Specifications

Yellow
12148-71-9
Powder
C18H30Ir2O2
MFCD08459360
Light, air, moisture sensitive
1g
Ir nominally 58%
Ir nominally 58%
662.88
1, 5-Cyclooctadiene(methoxy)iridium(I) dimer
Methoxy(cyclooctadiene)iridium(I) dimer

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit