Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetic acid, 97%

Catalog No. AAB229170B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB229170B 1000 g
AAB2291718 50 g
AAB2291730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB229170B Supplier Thermo Scientific Chemicals Supplier No. B229170B
Only null left
Add to Cart

CAS: 625-45-6 | C3H6O3 | 90.078 g/mol

Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00004308
InChI Key RMIODHQZRUFFFF-UHFFFAOYSA-N
Synonym methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
PubChem CID 12251
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O

Specifications

Melting Point 8°C
Density 1.174
pH 1.6
Boiling Point 202°C to 204°C
Flash Point 114°C (237°F)
Odor Odorless
Refractive Index 1.4165
Linear Formula CH3OCH2CO2H
Quantity 1000 g
UN Number UN3265
Beilstein 635800
Solubility Information Miscible with water.
Formula Weight 90.08
Percent Purity 97%
Chemical Name or Material Methoxyacetic acid
Show More Show Less
Hazard Category H302-H314-H335-H360FD
Hazard Statement GHS H Statement
H360-H314-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-894-6
RTECSNumber AI8650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.