Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetonitrile, 98%

Catalog No. AC125660250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125660250 25 g Glass bottle
AC125660050 5 g Glass bottle
AC125661000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC125660250 Supplier Thermo Scientific Chemicals Supplier No. 125660250
Add to Cart
This item is not returnable. View return policy

CAS: 1738-36-9 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-36-9
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00001892
InChI Key QKPVEISEHYYHRH-UHFFFAOYSA-N
PubChem CID 74442
IUPAC Name 2-methoxyacetonitrile
SMILES COCC#N

Specifications

Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 118.0°C to 119.0°C (731.0 mmHg)
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3800 to 1.3830 (20°C, 589nm)
Linear Formula CH3OCH2CN
Quantity 25 g
Beilstein 03,242
Fieser 05,436
Specific Gravity 0.95
Synonym methoxyacetonitrile, acetonitrile, methoxy, acetonitrile, 2-methoxy, ncch2och3, 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile,2-methoxy, intermediates-zcf02294, acmc-209e7f
Formula Weight 71.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methoxyacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
Wash face,hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 217-092-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.