Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

Catalog No. p-7023172
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1779-58-4 | C21H20BrO2P | 415.27 g/mol

(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-58-4
Molecular Formula C21H20BrO2P
Molecular Weight (g/mol) 415.27
MDL Number MFCD00011801
InChI Key VCWBQLMDSMSVRL-UHFFFAOYSA-M
Synonym 2-methoxy-2-oxoethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenylphosphonium bromide, carbomethoxymethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenyl phosphonium bromide, 2-methoxy-2-oxoethyl triphenylphosphanium bromide, 2-methoxy-2-oxoethyl-triphenylphosphanium bromide, 2-methoxy-2-oxo-ethyl-triphenyl-phosphonium bromide, phosphonium, 2-methoxy-2-oxoethyl triphenyl-, bromide, acmc-209edx
PubChem CID 2733211
IUPAC Name (2-methoxy-2-oxoethyl)-triphenylphosphanium;bromide
SMILES [Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼163°C (decomposition)
Quantity 25 g
Beilstein 3812975
Sensitivity Hygroscopic
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 415.28
Percent Purity ≥98%
Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-222-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.