Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methoxychlor

Click to view available options
Quantity:
5g
25g
72-43-5
C16H15Cl3O2
345.644
MFCD00000803
IAKOZHOLGAGEJT-UHFFFAOYSA-N
methoxychlor,p,p'-methoxychlor,dmdt,dimethoxy-ddt,methoxy-ddt,methoxcide,metox,maralate,marlate,moxie
4115
CHEBI:6842
1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl
72-43-5
C16H15Cl3O2
345.644
MFCD00000803
IAKOZHOLGAGEJT-UHFFFAOYSA-N
methoxychlor,p,p'-methoxychlor,dmdt,dimethoxy-ddt,methoxy-ddt,methoxcide,metox,maralate,marlate,moxie
4115
CHEBI:6842
1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Molecule with estrogenic activity

Color White
Melting Point 87°C
Quantity 5g
UN Number UN3077
Beilstein 2057367
Merck Index 14,5990
Formula Weight 345.65
Physical Form Crystalline Powder
Chemical Name or Material Methoxychlor

GHS H Statement
H351-H302-H312-H332
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : KJ3675000