Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%, Thermo Scientific™

Catalog No. AC210560250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Catalog No. AC210560250

Supplier: Thermo Scientific Chemicals 210560250

Only null left
Add to Cart

Chemical Identifiers

CAS 3939-01-3
Molecular Formula C14H17NO3·HCl
Molecular Weight (g/mol) 283.76
MDL Number MFCD00012799
InChI Key BRADBAOVPACOQQ-UHFFFAOYNA-N
Synonym methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate, hcl, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5
PubChem CID 107479

Specifications

Melting Point 172°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 283.76
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%
Hazard Category HYGROSCOPIC
EINECSNumber 223-522-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.