Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-cyclohexene-1-carboxylate, 98%

Catalog No. p-2691693
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 18448-47-0 | C8H12O2 | 140.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18448-47-0
MDL Number MFCD00001544
InChI Key KXPWRCPEMHIZGU-UHFFFAOYSA-N
Synonym methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene
PubChem CID 87647
IUPAC Name methyl cyclohexene-1-carboxylate
SMILES COC(=O)C1=CCCCC1

Specifications

Color Colorless
Density 1.0300g/mL
Boiling Point 190°C to 192°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H12O2
Refractive Index 1.4755 to 1.4775
Linear Formula C6H9CO2CH3
Quantity 5 g
Beilstein 09, IV, 112
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Molecular Weight (g/mol) 140.18
Formula Weight 140.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 1-cyclohexene-1-carboxylate
Show More Show Less
EINECSNumber 242-331-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.