Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate, 97%, Thermo Scientific™

Catalog No. p-7055086
missing translation for 'changeView'
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63577MD 250 mg
AAH6357703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH63577MD Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' H63577MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59857-86-2
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00015898
InChI Key JFZRRXGXEHUJCI-UHFFFAOYNA-N
PubChem CID 108402
IUPAC Name methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
SMILES COC(=O)C1CN(C)C(=O)C1

Specifications

Quantity 250 mg
Synonym methyl 1-methyl-2-oxopyrrolidine-4-carboxylate, 1-methyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester, 3-pyrrolidinecarboxylic acid, 1-methyl-5-oxo-, methyl ester, 1-methyl-4-methyloxycarbonyl-2-pyrrolidone, 1-methyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester, 5-22-06-00040 beilstein handbook reference, 4-carbomethoxy-1-methyl-pyrrolidone, methyl n-methyl-2-pyrrolidone-4-carboxylate, methyl1-methyl-2-oxopyrrolidine-4-carboxylate, methyl 1-methyl-5-oxo-3-pyrrolidinecarboxylate
Formula Weight 157.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UY0557000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.