Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 10-undecynoate, 96%, Thermo Scientific™

Catalog No. aah5345306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH5345306

Supplier: Thermo Scientific Chemicals H5345306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2777-66-4
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00190208
InChI Key TXFNJLOAXFOJGK-UHFFFAOYSA-N
Synonym methyl 10-undecynoate, 10-undecynoic acid, methyl ester, 10-undecynoic acid,methyl ester
PubChem CID 554128
IUPAC Name methyl undec-10-ynoate
SMILES COC(=O)CCCCCCCCC#C

Specifications

Density 0.908
Boiling Point 124°C (10 mmHg)
Refractive Index 1.4464
Linear Formula HC≡C(CH2)8CO2CH3
Quantity 5 g
Formula Weight 196.29
Percent Purity 96%
Chemical Name or Material Methyl 10-undecynoate
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.