Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1H-pyrazole-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7032619
Click to view available options
Quantity:
1 g

Methyl 1H-pyrazole-3-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1H-pyrazole-3-carboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15366-34-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00649381
InChI Key ORUCTBNNYKZMSK-UHFFFAOYSA-N
Synonym methyl 1h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid methyl ester, methyl pyrazole-3-carboxylate, 2h-pyrazole-3-carboxylic acid methyl ester, 3-methoxycarbonylpyrazole, pyrazole-3-carboxylic acid methyl ester, methyl 2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, methyl ester, pubchem14332, 5-methoxycarbonylpyrazole
PubChem CID 565662
IUPAC Name methyl 1H-pyrazole-5-carboxylate
SMILES COC(=O)C1=CC=NN1

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Methyl 1H-pyrazole-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.