Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3,6-Tri-O-benzoyl-alpha-D-galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. M193325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M193325G 25 g
M19335G 5 g
M1933250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M193325G Supplier TCI America Supplier No. M193325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3601-36-3
Molecular Formula C28H26O9
Molecular Weight (g/mol) 506.507
MDL Number MFCD06200842
InChI Key WXFFEILSURAFKL-SUWSLWCISA-N
Synonym methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
PubChem CID 10929259
IUPAC Name [(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
SMILES COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 143°C
Color White
Quantity 25 g
Formula Weight 506.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,6-Tri-O-benzoyl-alpha-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.