Learn More
Methyl 2,3-dichloropropionate, 98%
Description
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.
Solubility
Soluble in ether, acetone, ethanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
Specifications
Specifications
| Density | 1.326 |
| Boiling Point | 91°C to 92°C (50 mmHg) |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.453 |
| Quantity | 100 g |
| UN Number | UN3265 |
| Beilstein | 1701891 |
| Synonym | methyl 2,3-dichloropropionate, propanoic acid, 2,3-dichloro-, methyl ester, 2,3-dichloropropionic acid methyl ester, methyl-2,3-dichloropropionate, methyl alpha,beta-dichloropropionate, methyl .alpha.,.beta.-dichloropropionate, propionic acid, 2,3-dichloro-, methyl ester, propanoic acid,3-dichloro-, methyl ester, propionic acid,3-dichloro-, methyl ester, methyl-2,3-dichloropropioate |
| Solubility Information | Soluble in ether,acetone,ethanol. |
| SMILES | COC(=O)C(CCl)Cl |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 222-940-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.