Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4,5-trichlorophenyl Sulfide 97%, Thermo Scientific™

Catalog No. AC181750100
Click to view available options
Quantity:
10 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-78-4
Molecular Formula C7H5Cl3S
Molecular Weight (g/mol) 227.527
InChI Key RWDHTEMSFGMSDP-UHFFFAOYSA-N
Synonym 2,4,5-trichlorothioanisole, methyl 2,4,5-trichlorophenyl sulfide, unii-7a1g9vo987, 1,2,4-trichloro-5-methylsulfanyl benzene, methyl 2,4,5-trichlorophenyl sulphide, acmc-1afqu, benzene, 1,2,4-trichloro-5-methylthio, 2,4,5-trichlorophenyl methyl sulfide, methyl 2,4,5-trichlorophenyl sulfane, 1,2,5-trichloro-4-methylthio benzene
PubChem CID 77819
IUPAC Name 1,2,4-trichloro-5-methylsulfanylbenzene
SMILES CSC1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Linear Formula Cl3C6H2SCH3
Quantity 10 g
Formula Weight 227.54
Chemical Name or Material Methyl 2, 4, 5-trichlorophenyl sulfide, 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.