Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dihydroxybenzoate, 97%

Catalog No. p-7027755
Click to view available options
Quantity:
25 g
100 g

CAS: 2150-47-2 | C8H8O4 | 168.148 g/mol

Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-47-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002276
InChI Key IIFCLXHRIYTHPV-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate
PubChem CID 16523
IUPAC Name methyl 2,4-dihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)O

Specifications

Melting Point 116°C to 119°C
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dihydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-428-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.