Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dioxo-4-(2-thienyl)butyrate, 97%, Thermo Scientific™

Catalog No. AAH6139603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6139603 1 g
AAH61396MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6139603 Supplier Thermo Scientific Chemicals Supplier No. H6139603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57409-51-5
Molecular Formula C9H8O4S
Molecular Weight (g/mol) 212.22
MDL Number MFCD01249731
InChI Key TZNHHDJRXUBHRB-ALCCZGGFSA-N
Synonym methyl 2,4-dioxo-4-thiophen-2-yl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid methyl ester, methyl 2,4-dioxo-4-thien-2-ylbutanoate, methyl 2,4-dioxo-4-2-thienyl butanoate, methyl 2,4-dioxo-4-2-thienyl butyrate, methyl 2-thienoylpyruvate, methyl 4-2-thienyl-2,4-dioxobutanoate, methyl 2,4-dioxo-4-thien-2-yl butanoate, methyl2,4-dioxo-4-thiophen-2-yl butanoate, 2-thiophenebutanoicacid, a,g-dioxo-, methyl ester
PubChem CID 2771673
IUPAC Name methyl 2,4-dioxo-4-thiophen-2-ylbutanoate
SMILES COC(=O)C(\O)=C\C(=O)C1=CC=CS1

Specifications

Quantity 1 g
UN Number UN1325
Formula Weight 212.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2,4-dioxo-4-(2-thienyl)butyrate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.