Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,5-dihydroxybenzoate, 98%

Catalog No. AAA1040714
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1040714

Supplier: Thermo Scientific Chemicals A1040714

Only null left
Add to Cart

CAS: 2150-46-1 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-46-1
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016464
InChI Key XGDPKUKRQHHZTH-UHFFFAOYSA-N
Synonym methyl gentisate, gentisic acid methyl ester, methyl2,5-dihydroxybenzoate, benzoic acid, 2,5-dihydroxy-, methyl ester, methyl dihydroxybenzoate, 2,5-dihydroxybenzoic acid methyl ester, gentisic acid, methyl ester, methyl-2,5-dihydroxybenzoate, unii-ls28b004km, rarechem al bf 0043
PubChem CID 75077
IUPAC Name methyl 2,5-dihydroxybenzoate
SMILES COC(=O)C1=CC(O)=CC=C1O

Specifications

Melting Point 86°C to 87°C
Quantity 25 g
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material Methyl 2,5-dihydroxybenzoate
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 218-427-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.