Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-amino-5-bromobenzoate, 98%

Catalog No. AAA1666614
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 52727-57-8 | C8H8BrNO2 | 230.06 g/mol

Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

Solubility
Soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents, strong reducing agents, acid halides, acid anhydrides, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52727-57-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00144761
InChI Key QVNYNHCNNGKULA-UHFFFAOYSA-N
Synonym methyl 5-bromoanthranilate, 2-amino-5-bromobenzoic acid methyl ester, 5-bromoanthranilic acid methyl ester, 2-amino-5-bromo-benzoic acid methyl ester, methyl 5-bromo-2-aminobenzoate, benzoic acid, 2-amino-5-bromo-, methyl ester, benzoic acid, 2-amino-5-bromo-, methyl ester, hydrochloride, pubchem3344, methyl-5-bromoanthranilate, acmc-1ats7
PubChem CID 736224
IUPAC Name methyl 2-amino-5-bromobenzoate
SMILES COC(=O)C1=CC(Br)=CC=C1N

Specifications

Melting Point 71°C to 73°C
Quantity 25 g
Beilstein 972720
Solubility Information Soluble in water.
Formula Weight 230.06
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-bromobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.