Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromo-3-fluoropropionate, 97%

Catalog No. AAH3179003
Click to view available options
Quantity:
250 mg
1 g

CAS: 1537-52-6 | C4H6BrFO2 | 184.99 g/mol

In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1537-52-6
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.99
MDL Number MFCD09800641
InChI Key BXTYDNLOFHBZFS-UHFFFAOYNA-N
Synonym 2-bromo-3-fluoropropionic acid methyl ester, methyl 2-bromo-3-fluoropropionate, methyl 2-bromo-3-fluoro-propanoate, methyl alpha-bromo-beta-fluoropropionate, methyl 2-bromo-3-fluoroproprionate, propanoic acid, 2-bromo-3-fluoro-, methyl ester
PubChem CID 5271584
IUPAC Name methyl 2-bromo-3-fluoropropanoate
SMILES COC(=O)C(Br)CF

Specifications

Boiling Point 50°C to 54°C (8 mmHg)
Quantity 1 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 184.99
Percent Purity 97%
Chemical Name or Material Methyl 2-bromo-3-fluoropropionate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.