Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromo-5-methoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M193425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M193425G 25 g
M19345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M193425G Supplier TCI America Supplier No. M193425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35450-36-3
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00051594
InChI Key VRTQLDFCPNVQNT-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxybenzoic acid methyl ester, 2-bromo-5-methoxy-benzoic acid methyl ester, methyl-2-bromo-5-methoxybenzoate, benzoic acid, 2-bromo-5-methoxy-, methyl ester, pubchem3965, acmc-1ahhz, methyl 6-bromo-m-anisate, rarechem al bf 1111, attercop-chm at108617, methyl 2-bromo-5-methoxylbenzoate
PubChem CID 2776849
IUPAC Name methyl 2-bromo-5-methoxybenzoate
SMILES COC(=O)C1=CC(OC)=CC=C1Br

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromo-5-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.