Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromobenzoate, 99%

Catalog No. AAA1243522
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 610-94-6 | C8H7BrO2 | 215.05 g/mol

Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016328
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
PubChem CID 11894
IUPAC Name methyl 2-bromobenzoate
SMILES COC(=O)C1=CC=CC=C1Br

Specifications

Density 1.532
Boiling Point 252°C
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 100 g
Beilstein 1862501
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material Methyl 2-bromobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-241-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.