Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals

Catalog No. AAH6354103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6354103 1 g
AAH6354106 5 g
AAH63541MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6354103 Supplier Thermo Scientific Chemicals Supplier No. H6354103

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170235-26-4
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD06659908
InChI Key YOWKNNKTQWCYNC-UHFFFAOYSA-N
PubChem CID 2763213
IUPAC Name methyl 2-bromo-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)Br

Specifications

Quantity 1 g
Synonym methyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid methyl ester, 2-bromo-4-methoxycarbonyl-1,3-thiazole, methyl2-bromothiazole-4-carboxylate, methyl-2-bromothiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-4-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-4-carboxylic acid methyl ester, pubchem8920, acmc-1c5qt
Solubility Information Slightly soluble in water.
Formula Weight 222.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 2-bromothiazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.