Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chlorobenzoate, 98%

Catalog No. AAA1123722
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 610-96-8 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-96-8
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00016337
InChI Key JAVRNIFMYIJXIE-UHFFFAOYSA-N
Synonym methyl o-chlorobenzoate, methyl-2-chlorobenzoate, benzoic acid, 2-chloro-, methyl ester, 2-chlorobenzoic acid methyl ester, unii-g344nes07r, benzoic acid, o-chloro-, methyl ester, benzoicacid, 2-chloro-, methyl ester, pubchem3707, methyl-o-chlorobenzoate, acmc-209moz
PubChem CID 11895
IUPAC Name methyl 2-chlorobenzoate
SMILES COC(=O)C1=CC=CC=C1Cl

Specifications

Density 1.191
Boiling Point 233°C to 235°C
Flash Point 108°C (226°F)
Refractive Index 1.536
Quantity 100 g
Beilstein 1364684
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material Methyl 2-chlorobenzoate
EINECSNumber 210-242-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.