Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chloropropionate, 97%

Catalog No. AAA185410I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA185410I 5000 g
AAA1854130 250 g
AAA185410B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA185410I Supplier Thermo Scientific Chemicals Supplier No. A185410I

CAS: 17639-93-9 | C4H7ClO2 | 122.55 g/mol

Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

Solubility
Insoluble in water

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17639-93-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00000863
InChI Key JLEJCNOTNLZCHQ-UHFFFAOYNA-N
Synonym methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
PubChem CID 28659
IUPAC Name methyl 2-chloropropanoate
SMILES COC(=O)C(C)Cl

Specifications

Melting Point <-10°C
Boiling Point 132°C to 133°C
Flash Point 38°C (100°F)
Quantity 5000 g
UN Number UN2933
Beilstein 1421535
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material Methyl 2-chloropropionate
Hazard Category Warning
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL 2-CHLOROPROPIONATE
RTECSNumber UE9100000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.