Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Chloropropionate 95.0+%, TCI America™
SDP

Catalog No. C0970100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0970100ML 100 mL
C097025ML 25 mL
C0970500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0970100ML Supplier TCI America Supplier No. C0970100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17639-93-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00000863
InChI Key JLEJCNOTNLZCHQ-UHFFFAOYNA-N
Synonym methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
PubChem CID 28659
IUPAC Name methyl 2-chloropropanoate
SMILES COC(=O)C(C)Cl

Specifications

Color Colorless
Boiling Point 132°C
Quantity 100 mL
UN Number 2933
Formula Weight 122.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Chloropropionate
EINECSNumber (2)-3084
RTECSNumber UE9100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.