Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Fluoro-2-methylpropionate 95.0+%, TCI America™
SDP

Catalog No. M282725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M282725G 25 g
M28275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M282725G Supplier TCI America Supplier No. M282725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 338-76-1
Molecular Formula C5H9FO2
Molecular Weight (g/mol) 120.123
MDL Number MFCD03265444
InChI Key OEGBOFOVYSOERL-UHFFFAOYSA-N
Synonym 2-Fluoro-2-methylpropionic Acid Methyl Ester
PubChem CID 9793744
IUPAC Name methyl 2-fluoro-2-methylpropanoate
SMILES CC(C)(C(=O)OC)F

Specifications

Color Colorless
Boiling Point 109°C
Quantity 25 g
UN Number 2924
Formula Weight 120.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Fluoro-2-methylpropionate
EINECSNumber (2)-3792
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.