Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-hydroxy-5-nitrobenzoate, 98%

Catalog No. p-7028136
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 17302-46-4 | C8H7NO5 | 197.146 g/mol

Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17302-46-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00024695
InChI Key UUBFELFUKFJSRD-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-5-nitrobenzoate, methyl 5-nitrosalicylate, methyl-5-nitrosalicylate, 5-nitrosalicylic acid methyl ester, benzoic acid, 2-hydroxy-5-nitro-, methyl ester, 2-hydroxy-5-nitrobenzoic acid methyl ester, methyl 5-nitro-salicylate, acmc-20a62q, methyl 5-nitro-2-hydroxybenzoate, methyl-2-hydroxy-5-nitrobenzoate
PubChem CID 87041
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

Melting Point 114°C to 117°C
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-5-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-330-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.