Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-hydroxyisobutyrate, 99%

Catalog No. p-164240
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle

CAS: 2110-78-3 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2110-78-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004457
InChI Key XYVQFUJDGOBPQI-UHFFFAOYSA-N
Synonym methyl 2-hydroxyisobutyrate, methyl 2-methyllactate, 2-hydroxyisobutyric acid methyl ester, methyl 2-hydroxy-2-methylpropionate, methyl2-hydroxyisobutyrate, propanoic acid, 2-hydroxy-2-methyl-, methyl ester, methyl alpha-hydroxyisobutyrate, methyl .alpha.-hydroxyisobutyrate, methyl ester of 2-methyllactic acid, methyl 2-hydroxy-2-methyl-propanoate
PubChem CID 75024
IUPAC Name methyl 2-hydroxy-2-methylpropanoate
SMILES CC(C)(C(=O)OC)O

Specifications

Color Colorless
Density 1.0300g/mL
Boiling Point 137°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4100 to 1.412
Linear Formula (CH3)2C(OH)CO2CH3
Quantity 5 g
Beilstein 03, II, 223
Specific Gravity 1.03
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2-hydroxyisobutyrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-301-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.