Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-iodobenzoate, 98%

Catalog No. AAA1320430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320430 250 g
AAA1320409 10 g
AAA1320418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1320430 Supplier Thermo Scientific Chemicals Supplier No. A1320430

CAS: 610-97-9 | C8H7IO2 | 262.046 g/mol

2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-97-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00016351
InChI Key BXXLTVBTDZXPTN-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0
PubChem CID 69132
IUPAC Name methyl 2-iodobenzoate
SMILES COC(=O)C1=CC=CC=C1I

Specifications

Density 1.73
Boiling Point 274°C
Flash Point >110°C (230°F)
Refractive Index 1.604
Quantity 250 g
Beilstein 2206859
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material Methyl 2-iodobenzoate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-243-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.