Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylhexanoic acid, 98%

Catalog No. AC457411000
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 628-46-6 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-46-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00036494
InChI Key MHPUGCYGQWGLJL-UHFFFAOYSA-N
Synonym isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
PubChem CID 12344
IUPAC Name 5-methylhexanoic acid
SMILES CC(C)CCCC(O)=O

Specifications

Density 0.9050g/mL
Boiling Point 121.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH(CH3)(CH2)3CO2H
Quantity 100 g
Specific Gravity 0.905
Solubility Information Solubility in water: slightly. Other solubilities: soluble in organic solvents
Formula Weight 130.18
Percent Purity 98%
Chemical Name or Material 5-Methylhexanoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.