Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-nitrobenzoate, 98+%

Catalog No. p-7026788
Click to view available options
Quantity:
25 g
100 g

CAS: 606-27-9 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-27-9
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007136
InChI Key AOXPHVNMBPFOFS-UHFFFAOYSA-N
Synonym methyl o-nitrobenzoate, benzoic acid, 2-nitro-, methyl ester, 2-nitro-benzoic acid methyl ester, benzoic acid, o-nitro-, methyl ester, 2-nitrobenzoic acid methyl ester, methyl-2-nitrobenzoate, methyl o-nitro benzoate, methyl ortho-nitrobenzoate, acmc-209ml4, nitrobenzoic acid methyl ester
PubChem CID 69072
IUPAC Name methyl 2-nitrobenzoate
SMILES COC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -13°C
Density 1.285
Boiling Point 275°C
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 25 g
Beilstein 2050101
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 2-nitrobenzoate
EINECSNumber 210-111-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.