Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-oxocyclohexanecarboxylate, 90%

Catalog No. p-3523491
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle

CAS: 41302-34-5 | C8H12O3 | 156.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41302-34-5
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.2
MDL Number MFCD00077699
InChI Key JEENWEAPRWGXSG-UHFFFAOYSA-N
Synonym methyl 2-oxocyclohexanecarboxylate, 2-methoxycarbonylcyclohexanone, 2-carbomethoxycyclohexanone, cyclohexanecarboxylic acid, 2-oxo-, methyl ester, methyl cyclohexanone-2-carboxylate, 2-oxo-cyclohexanecarboxylic acid methyl ester, 2-hydroxy methyl cyclohexanone, 2-oxocyclohexanecarboxylic acid, methyl ester, pubchem20380, acmc-1ao3u
PubChem CID 170489
IUPAC Name methyl 2-oxocyclohexane-1-carboxylate
SMILES COC(=O)C1CCCCC1=O

Specifications

Color Colorless to Yellow
Density 1.1090g/mL
Boiling Point 50°C (0.2 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. may occur as tautomers (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.486
Quantity 10 mL
Beilstein 10, IV, 2606
Specific Gravity 1.109
Formula Weight 156.2
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl 2-oxocyclohexanecarboxylate
Show More Show Less
EINECSNumber 255-306-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.