Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(p-Tolyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. M180925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M180925G 25 g
M18095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M180925G Supplier TCI America Supplier No. M180925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 114772-34-8
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD03453661
InChI Key IHNIAWHITVGYJJ-UHFFFAOYSA-N
PubChem CID 1393873
IUPAC Name methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate
SMILES COC(=O)C1=CC=CC=C1C1=CC=C(C)C=C1

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 25 g
Synonym 2-(p-Tolyl)benzoic Acid Methyl Ester, Methyl 2-(4-Methylphenyl)benzoate, Methyl 4′-Methylbiphenyl-2-carboxylate, 4′-Methylbiphenyl-2-carboxylic Acid Methyl Ester
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(p-Tolyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.