Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(2-hydroxyphenyl)propionate, 97%

Catalog No. p-7081994
Click to view available options
Quantity:
5 g
25 g

CAS: 20349-89-7 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20349-89-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00067757
InChI Key YHXYRISRGHSPNV-UHFFFAOYSA-N
Synonym methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester
PubChem CID 2794569
IUPAC Name methyl 3-(2-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1O

Specifications

Melting Point 40°C to 44°C
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material Methyl 3- (2-hydroxyphenyl)propionate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.