Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%, Thermo Scientific™

Catalog No. AAH5577003
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474334-89-9
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD07369792
InChI Key LPOJJZXQESTVEV-UHFFFAOYSA-N
Synonym methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
PubChem CID 16217966
IUPAC Name methyl 3-(4-methylpiperazin-1-yl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1

Specifications

Color Colorless to Yellow
Density 1.119 g/mL
Boiling Point 169°C to 172°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 1 g
Formula Weight 234.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3-(4-methyl-1-piperazinyl)benzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.