Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-aminobenzoate, 98%

Catalog No. aaa1476606
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 4518-10-9 | C8H9NO2 | 151.17 g/mol

Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water (0.1-1%) .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4518-10-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017102
InChI Key VZDNXXPBYLGWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-, methyl ester, methyl-3-aminobenzoate, 3-aminobenzoic acid methyl ester, m-aminobenzoic acid, methyl ester, 3-amino-benzoic acid methyl ester, methyl m-aminobenzoate, methyl3-aminobenzoate, methyl-m-aminobenzoate, pubchem16709, acmc-1aeif
PubChem CID 78274
IUPAC Name methyl 3-aminobenzoate
SMILES COC(=O)C1=CC(=CC=C1)N

Specifications

Melting Point 49°C to 53°C
Density 1.232
Quantity 5 g
Beilstein 1101468
Solubility Information Slightly soluble in water (0.1-1%) .
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material Methyl 3-aminobenzoate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-842-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.