Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-aminopyridine-4-carboxylate, 97%

Catalog No. p-7032518
Click to view available options
Quantity:
1 g
10 g

CAS: 55279-30-6 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55279-30-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD02082535
InChI Key XLQIGLBALJNHKR-UHFFFAOYSA-N
PubChem CID 1488824
IUPAC Name methyl 3-aminopyridine-4-carboxylate
SMILES COC(=O)C1=C(C=NC=C1)N

Specifications

Melting Point 84°C to 86°C
Quantity 1 g
Synonym methyl 3-aminoisonicotinate, 3-amino-isonicotinic acid methyl ester, 3-aminopyridine-4-carboxylic acid methyl ester, methyl3-aminoisonicotinate, 3-aminoisonicotinic acid methyl ester, 4-pyridinecarboxylic acid, 3-amino-, methyl ester, 3-amino-4-pyridinecarboxylic acid methyl ester, pubchem9760, acmc-1avm8, methyl 3-amino isonicotinate
Formula Weight 152.2
Percent Purity 97%
Chemical Name or Material Methyl 3-aminopyridine-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.