Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(Bromomethyl)benzoate 97.0+%, TCI America™
SDP

Catalog No. B222425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B222425G 25 g
B22245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B222425G Supplier TCI America Supplier No. B222425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1129-28-8
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00051437
InChI Key YUHSMQQNPRLEEJ-UHFFFAOYSA-N
PubChem CID 517981
IUPAC Name methyl 3-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=CC(CBr)=C1

Specifications

Melting Point 44°C
Boiling Point 169°C
Quantity 25 g
UN Number 3261
Synonym methyl 3-bromomethyl benzoate, m-carbomethoxybenzyl bromide, 3-bromomethyl benzoic acid methyl ester, benzoic acid, 3-bromomethyl-, methyl ester, methyl-3-bromomethylbenzoate, methyl alpha-bromo-m-toluate, methyl3-bromomethyl benzoate, methyl-3-bromomethyl benzoate, 3-carbomethoxybenzyl bromide
Formula Weight 229.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-(Bromomethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.