Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Bromopropionate 99.0+%, TCI America™
SDP

Catalog No. B0648100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0648100G 100 g
B064825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0648100G Supplier TCI America Supplier No. B0648100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3395-91-3
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000250
InChI Key KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Synonym methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
PubChem CID 76934
IUPAC Name methyl 3-bromopropanoate
SMILES COC(=O)CCBr

Specifications

Color Colorless
Boiling Point 105°C
Quantity 100 g
Formula Weight 167.00
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromopropionate
RTECSNumber UA2451790
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.