Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-cyanoindole-6-carboxylate, 97%

Catalog No. AAH6310006
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6310006 5 g
AAH63100MD 250 mg
AAH6310003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6310006 Supplier Thermo Scientific Chemicals Supplier No. H6310006
Add to Cart

CAS: 1000576-51-1 | C11H8N2O2 | 200.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000576-51-1
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD09878564
InChI Key YBZVEPCMUWGFNW-UHFFFAOYSA-N
Synonym methyl 3-cyanoindole-6-carboxylate, 3-cyano-1h-indole-6-carboxylic acid methyl ester, 1h-indole-6-carboxylicacid, 3-cyano-, methyl ester, 1h-indole-6-carboxylic acid, 3-cyano-, methyl ester
PubChem CID 37819073
IUPAC Name methyl 3-cyano-1H-indole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C(=CN2)C#N

Specifications

Quantity 5 g
Formula Weight 200.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-cyanoindole-6-carboxylate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.