Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-cyclohexenecarboxylate, 97%, Thermo Scientific™

Catalog No. AC452860250
Click to view available options
Quantity:
25 mL
100 mL

Chemical Identifiers

CAS 6493-77-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00053266
InChI Key IPUNVLFESXFVFH-UHFFFAOYNA-N
Synonym methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
PubChem CID 96926
IUPAC Name methyl cyclohex-3-ene-1-carboxylate
SMILES COC(=O)C1CCC=CC1

Specifications

Density 1.0200g/mL
Boiling Point 185.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 mL
Specific Gravity 1.02
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclohexenecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 229-376-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.