Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-N,N-Dimethylaminoazobenzene-4'-isothiocyanate, Thermo Scientific™

Catalog No. AC241892500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7612-98-8
Molecular Formula C15H14N4S
Molecular Weight (g/mol) 282.37
MDL Number MFCD00004812
InChI Key OSWZKAVBSQAVFI-UHFFFAOYSA-N
Synonym dabitc, 4-n,n-dimethylamino azobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenylazo-n,n-dimethylaniline, 4-dimethylaminoazobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenyl azo-n,n-dimethylaniline, 4-dimethylamino azobenzene 4'-isothiocyanate, 4-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-e-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-n,n-dimethylaminoazobenzene-4'-isothiocyanate, 4-dimethylamino-4'-isothiocyanatoazobenzene
PubChem CID 82084
IUPAC Name 4-[2-(4-isothiocyanatophenyl)diazen-1-yl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 167°C to 171°C
Color Rust
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 282.35
Percent Purity ≥97.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-N, N-Dimethylaminoazobenzene-4'-isothiocyanate
Hazard Category MOISTURE SENSITIVE
EINECSNumber 231-521-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.