Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
113212-14-9
C7H10N2O2
154.17
FREGRQYTSPAJDX-AATRIKPKSA-N
3-dimethylamino-2-isocyanoacrylic acid methyl ester,methyl e-3-dimethylamino-2-isocyanoprop-2-enoate,methyl 2e-3-dimethylamino-2-isocyanoprop-2-enoate
6387542
methyl (E)-3-(dimethylamino)-2-isocyanoprop-2-enoate
CN(C)C=C(C(=O)OC)[N+]#[C-]
113212-14-9
C7H10N2O2
154.17
FREGRQYTSPAJDX-AATRIKPKSA-N
3-dimethylamino-2-isocyanoacrylic acid methyl ester,methyl e-3-dimethylamino-2-isocyanoprop-2-enoate,methyl 2e-3-dimethylamino-2-isocyanoprop-2-enoate
6387542
methyl (E)-3-(dimethylamino)-2-isocyanoprop-2-enoate
CN(C)C=C(C(=O)OC)[N+]#[C-]
Melting Point 61.0°C to 67.0°C
Quantity 5g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 154.17
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material Methyl 3-(dimethylamino)-2-isocyanoacrylate
Show More Show Less

GHS Signal Word: Warning