Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-fluoro-4-hydroxybenzoate, 98%

Catalog No. aah61219md
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 403-01-0 | C8H7FO3 | 170.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-01-0
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00215838
InChI Key IYUSGKSCDUJSKS-UHFFFAOYSA-N
PubChem CID 7019368
IUPAC Name methyl 3-fluoro-4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C(F)=C1

Specifications

Quantity 250 mg
Synonym 3-fluoro-4-hydroxy-benzoic acid methyl ester, 3-fluoro-4-hydroxy benzoic acid methyl ester, 3-fluoro-4-hydroxybenzoic acid methyl ester, methyl3-fluoro-4-hydroxybenzoate, benzoic acid, 3-fluoro-4-hydroxy-, methyl ester, acmc-1ajcc, methyl 3-fluoro-4-hydroxy-benzoate, methyl 3-fluoro-4-hydroxybenzenecarboxylate, benzoic acid,3-fluoro-4-hydroxy-, methyl ester
Formula Weight 170.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 3-fluoro-4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.