Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Fluorophenylacetate 98.0+%, TCI America™
SDP

Catalog No. M253625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M253625G 25 g
M25365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M253625G Supplier TCI America Supplier No. M253625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64123-77-9
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD03093288
InChI Key OXYLCGRXCQARQV-UHFFFAOYSA-N
PubChem CID 2734108
IUPAC Name methyl 2-(3-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=CC(F)=C1

Specifications

Color Colorless
Quantity 25 g
Synonym methyl 2-3-fluorophenyl acetate, methyl 3-fluorophenylacetate, methyl3-fluorophenylacetate, methyl 3-fluorophenyl acetate, 3-fluorobenzeneacetic acid methyl ester, benzeneacetic acid, 3-fluoro-, methyl ester, 3-fluorophenylacetic acid methyl ester, 3-fluorophenyl acetic acid methyl ester, 3-fluoro-phenyl-acetic acid methyl ester, 2-3-fluorophenyl acetic acid methyl ester
Formula Weight 168.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Fluorophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.