Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-hydroxy-4-iodobenzoate, 97%, Thermo Scientific™

Catalog No. aah64354md
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157942-12-6
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD06203960
InChI Key LXCQVWRESZDFGW-UHFFFAOYSA-N
Synonym methyl-4-iodo-3-hydroxy benzoate, 3-hydroxy-4-iodobenzoic acid methyl ester, 3-hydroxy-4-iodo-benzoic acid methyl ester, methyl 3-hydroxy-4-iodobenzenecarboxylate, benzoic acid, 3-hydroxy-4-iodo-, methyl ester, rarechem al bf 0964, methyl 3-hydroxy4-iodobenzoate, methyl 4-iodo-3-hydroxybenzoate, methyl-3-hydroxy-4-iodobenzoate, methyl-4-iodo-3-hydroxybenzoate
PubChem CID 10378812
IUPAC Name methyl 3-hydroxy-4-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)O

Specifications

Quantity 250 mg
Beilstein 7289690
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 278.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-hydroxy-4-iodobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.