Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 3-hydroxythiophene-2-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
25g
5118-06-9
C6H6O3S
158.17
MFCD00055642
FYOPNADYSPZUAT-WAYWQWQTSA-N
methyl 3-hydroxy-2-thiophenecarboxylate,3-hydroxy-2-thiophenecarboxylic acid methyl ester,3-hydroxy-thiophene-2-carboxylic acid methyl ester,3-hydroxythiophene-2-carboxylic acid methyl ester,pubchem7744,acmc-1aqhj,maybridge3_005989,3-hydroxy-2-methoxycarbonylthiophene,3-hydroxy-2-methoxycarbonyl-thiophene,methyl 3-hydroxythiophen-2-caboxylate
581127
methyl 3-hydroxythiophene-2-carboxylate
COC(=O)C1=C(C=CS1)O
5118-06-9
C6H6O3S
158.17
MFCD00055642
FYOPNADYSPZUAT-WAYWQWQTSA-N
methyl 3-hydroxy-2-thiophenecarboxylate,3-hydroxy-2-thiophenecarboxylic acid methyl ester,3-hydroxy-thiophene-2-carboxylic acid methyl ester,3-hydroxythiophene-2-carboxylic acid methyl ester,pubchem7744,acmc-1aqhj,maybridge3_005989,3-hydroxy-2-methoxycarbonylthiophene,3-hydroxy-2-methoxycarbonyl-thiophene,methyl 3-hydroxythiophen-2-caboxylate
581127
methyl 3-hydroxythiophene-2-carboxylate
COC(=O)C1=C(C=CS1)O
Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 44°C to 48°C
Quantity 1g
Boiling Point 107°C to 109°C (13mmHg)
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate