Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-hydroxythiophene-2-carboxylate, 97%

Catalog No. p-7026231
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 5118-06-9 | C6H6O3S | 158.17 g/mol

Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5118-06-9
MDL Number MFCD00055642

Specifications

Melting Point 44°C to 48°C
Boiling Point 107°C to 109°C (13 mmHg)
Molecular Formula C6H6O3S
Quantity 1 g
Synonym methyl 3-hydroxy-2-thiophenecarboxylate, 3-hydroxy-2-thiophenecarboxylic acid methyl ester, 3-hydroxy-thiophene-2-carboxylic acid methyl ester, 3-hydroxythiophene-2-carboxylic acid methyl ester, pubchem7744, acmc-1aqhj, maybridge3_005989, 3-hydroxy-2-methoxycarbonylthiophene, 3-hydroxy-2-methoxycarbonyl-thiophene, methyl 3-hydroxythiophen-2-caboxylate
Solubility Information Slightly soluble in water.
InChI Key FYOPNADYSPZUAT-WAYWQWQTSA-N
SMILES COC(=O)C1=C(C=CS1)O
IUPAC Name methyl 3-hydroxythiophene-2-carboxylate
Molecular Weight (g/mol) 158.17
PubChem CID 581127
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.